Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FSIP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK092099.1 | - | 8 | 1625 |
Gene Symbol: HELLPAR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | JX088243.1 | - | 1 | 111498 |
Gene Symbol: TMEM45A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BX648832.1 | - | 2 | 2341 |
Gene Symbol: TNIP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011532257.2 | XP_011530559.1 | 14 | 5994 | ||
XM_017008619.1 | XP_016864108.1 | 16 | 6156 | |||
XM_017008620.1 | XP_016864109.1 | 16 | 6019 | |||
XM_017008621.1 | XP_016864110.1 | 15 | 6066 | |||
XM_017008622.1 | XP_016864111.1 | 15 | 6319 | |||
XM_017008623.1 | XP_016864112.1 | 15 | 5929 | |||
XM_017008624.1 | XP_016864113.1 | 14 | 5874 | |||
XM_017008625.1 | XP_016864114.1 | 13 | 6407 |