Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RAPGEF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006717067.3 | XP_006717130.1 | 1 | 60 | ||
XM_006717072.3 | XP_006717135.1 | 1 | 70 | |||
XM_006717074.3 | XP_006717137.1 | 1 | 60 | |||
XM_011518569.2 | XP_011516871.1 | 1 | 161 | |||
XM_011518570.2 | XP_011516872.1 | 1 | 53 | |||
XM_011518574.2 | XP_011516876.1 | 1 | 53 | |||
XM_011518575.2 | XP_011516877.1 | 1 | 53 | |||
XM_011518576.2 | XP_011516878.1 | 1 | 53 | |||
XM_011518577.2 | XP_011516879.1 | 1 | 53 | |||
XM_011518578.2 | XP_011516880.1 | 1 | 53 | |||
XM_011518579.2 | XP_011516881.1 | 1 | 53 | |||
XM_011518580.2 | XP_011516882.1 | 1 | 53 | |||
XM_011518581.2 | XP_011516883.1 | 1 | 53 | |||
XM_011518582.2 | XP_011516884.1 | 1 | 53 | |||
XM_017014637.1 | XP_016870126.1 | 1 | 53 | |||
XM_017014638.1 | XP_016870127.1 | 1 | 53 | |||
XM_017014639.1 | XP_016870128.1 | 1 | 53 | |||
XM_017014640.1 | XP_016870129.1 | 1 | 53 | |||
XM_017014641.1 | XP_016870130.1 | 1 | 53 | |||
GenBank mRNA | AF467069.1 | - | 1 | 905 |