Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LEF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK128255.1 | - | 2 | 1753 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | DA383766.1 | - | 1 | 494 | Chr. 4 - 108048830 - 108049463 [-] on Build GRCh38 |