Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GRTP1-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_046541.1 | 1 | 78 | |||
NR_120385.1 | 1 | 78 | ||||
GenBank mRNA | BC037976.1 | - | 1 | 78 |