Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RNPS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_005255049.3 | XP_005255106.1 | 1 | 1548 | ||
GenBank mRNA | AK021799.1 | - | 1 | 851 |