Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC105375531 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK000794.1 | - | 1 | 536 |
Gene Symbol: TBXAS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017012570.1 | XP_016868059.1 | 11 | 3709 |