Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PEX11A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271572.1 | NP_001258501.1 | 3 | 2053 | ||
NM_001271573.1 | NP_001258502.1 | 2 | 2030 | |||
NM_003847.2 | NP_003838.1 | 3 | 2146 | |||
GenBank mRNA | AL109728.1 | - | 1 | 258 | ||
AL360141.1 | - | 1 | 1260 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BQ437764.1 | - | 1 | 36 | Chr. 15 - 89681535 - 89682162 [-] on Build GRCh38 |