Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DERL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001304777.1 | NP_001291706.1 | 7 | 3163 | ||
NM_001304779.1 | NP_001291708.1 | 4 | 2802 | |||
NM_016041.4 | NP_057125.2 | 7 | 3166 | |||
NR_130905.1 | 6 | 3278 | ||||
NR_130906.1 | 6 | 3072 | ||||
GenBank mRNA | AK092218.1 | - | 1 | 932 |