Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AK4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005353.2 | NP_001005353.1 | 6 | 4800 | ||
NM_013410.3 | NP_037542.1 | 5 | 4683 | |||
NM_203464.2 | NP_982289.1 | 6 | 4689 | |||
XM_017000612.1 | XP_016856101.1 | 5 | 4435 | |||
XM_017000613.1 | XP_016856102.1 | 5 | 4621 | |||
GenBank mRNA | AK026966.1 | - | 1 | 320 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | CX788479.1 | - | 1 | 150 | Chr. 1 - 65229798 - 65230691 [+] on Build GRCh38 |