Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CTD-2201E9.1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_109935.1 | 3 | 1637 | |||
GenBank mRNA | AK057486.1 | - | 3 | 1637 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000510879.1 | - | 3 | 1637 | Chr. 5 - 9511333 - 9518086 [+] on Build GRCh38 |