Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UBE2V2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_005251300.2 | XP_005251357.1 | 4 | 2970 | ||
XM_011517583.2 | XP_011515885.1 | 4 | 2989 | |||
XM_017013808.1 | XP_016869297.1 | 4 | 2896 | |||
GenBank mRNA | AK123928.1 | - | 1 | 397 |