Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IST1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270975.1 | NP_001257904.1 | 10 | 2744 | ||
NM_001270976.1 | NP_001257905.1 | 11 | 2859 | |||
NM_001270977.1 | NP_001257906.1 | 9 | 2651 | |||
NM_001270978.1 | NP_001257907.1 | 7 | 2372 | |||
NM_001270979.1 | NP_001257908.1 | 6 | 2288 | |||
NM_014761.3 | NP_055576.2 | 10 | 2740 | |||
GenBank mRNA | BC038350.1 | - | 1 | 258 |