Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ENTPD1-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_134320.1 | 4 | 782 | |||
NR_134321.1 | 3 | 643 | ||||
NR_134322.1 | 4 | 786 | ||||
GenBank mRNA | BC028619.1 | - | 4 | 781 |
Gene Symbol: CC2D2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK022203.1 | - | 1 | 356 |