Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TUSC3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017013861.1 | XP_016869350.1 | 10 | 4951 | ||
GenBank mRNA | BX641112.1 | - | 1 | 3154 |