Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PCSK6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC043562.1 | - | 2 | 344 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BI766709.1 | - | 2 | 344 | Chr. 15 - 101295419 - 101303896 [+] on Build GRCh38 |