Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UCHL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017020726.1 | XP_016876215.1 | 9 | 1256 | ||
GenBank mRNA | AK093882.1 | - | 1 | 1348 |