Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PTPRE | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001323354.1 | NP_001310283.1 | 1 | 1508 | ||
GenBank mRNA | AK056111.1 | - | 1 | 1508 | ||
AL137560.1 | - | 1 | 1122 |