Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RAB3C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001317915.1 | NP_001304844.1 | 5 | 6741 | ||
NM_138453.3 | NP_612462.1 | 5 | 6703 | |||
GenBank mRNA | AK094685.1 | - | 1 | 750 |