Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NBPF25P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_104217.1 | 4 | 376 | |||
GenBank mRNA | AK125792.1 | - | 4 | 376 | ||
DB064097.1 | - | 3 | 454 |
Gene Symbol: LOC100996717 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017003047.1 | XP_016858536.1 | 6 | 846 | ||
XM_017003048.1 | XP_016858537.1 | 6 | 7541 | |||
XM_017003049.1 | XP_016858538.1 | 6 | 858 | |||
XM_017003050.1 | XP_016858539.1 | 6 | 1092 | |||
XM_017003052.1 | XP_016858541.1 | 7 | 992 |
Gene Symbol: LOC100996763 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011510256.2 | XP_011508558.1 | 6 | 1826 |
Gene Symbol: LOC101929796 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011542531.2 | XP_011540833.1 | 5 | 1030 | ||
XM_017003012.1 | XP_016858501.1 | 6 | 1125 | |||
XM_017003013.1 | XP_016858502.1 | 5 | 1030 | |||
XM_017003014.1 | XP_016858503.1 | 5 | 1030 |
Gene Symbol: NBPF14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011509395.2 | XP_011507697.1 | 2 | 264 |
Gene Symbol: NBPF19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017003019.1 | XP_016858508.1 | 1 | 37 |
Gene Symbol: NOTCH2NL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK075065.1 | - | 5 | 1017 |