Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FRY | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006719749.3 | XP_006719812.1 | 63 | 13961 | ||
XM_017020303.1 | XP_016875792.1 | 63 | 14438 | |||
XM_017020304.1 | XP_016875793.1 | 62 | 12862 | |||
XM_017020305.1 | XP_016875794.1 | 63 | 14457 | |||
XM_017020306.1 | XP_016875795.1 | 42 | 9599 | |||
XM_017020307.1 | XP_016875796.1 | 36 | 8952 | |||
GenBank mRNA | BC036296.1 | - | 1 | 519 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BG743355.1 | - | 1 | 518 | Chr. 13 - 32298257 - 32299084 [+] on Build GRCh38 |