Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ACER2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011517859.2 | XP_011516161.1 | 6 | 5005 | ||
GenBank mRNA | AK024898.1 | - | 1 | 1386 |