Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ORC4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001190879.2 | NP_001177808.1 | 15 | 4647 | ||
NM_001190881.2 | NP_001177810.1 | 11 | 4265 | |||
NM_001190882.2 | NP_001177811.1 | 13 | 4409 | |||
NM_002552.4 | NP_002543.2 | 14 | 4531 | |||
NM_181741.3 | NP_859525.1 | 14 | 4640 | |||
NM_181742.3 | NP_859526.1 | 14 | 4535 | |||
GenBank mRNA | AK128860.1 | - | 1 | 1862 |