Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PYHIN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_005244930.1 | XP_005244987.1 | 5 | 937 | ||
GenBank mRNA | AK024890.1 | - | 1 | 1088 | ||
AK308350.1 | - | 5 | 917 | |||
BC020822.1 | - | 5 | 916 |