Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DNASE1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC015002.1 | - | 1 | 577 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BM050845.1 | - | 1 | 419 | Chr. 16 - 3651995 - 3652897 [+] on Build GRCh38 |