Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ELAVL4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001324209.1 | NP_001311138.1 | 2 | 297 | ||
NM_001324212.1 | NP_001311141.1 | 2 | 297 | |||
NM_001324214.1 | NP_001311143.1 | 2 | 367 | |||
NR_136725.1 | 2 | 367 | ||||
XM_011540894.2 | XP_011539196.1 | 2 | 364 | |||
XM_011540895.2 | XP_011539197.1 | 2 | 268 | |||
GenBank mRNA | CK005107.1 | - | 2 | 300 | ||
S73887.1 | - | 1 | 27 |