Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HOXA3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_005249730.2 | XP_005249787.1 | 3 | 428 | ||
XM_006715715.2 | XP_006715778.1 | 3 | 428 | |||
XM_011515343.2 | XP_011513645.1 | 3 | 428 |