Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC100506679 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_125362.1 | 1 | 1007 | |||
GenBank mRNA | BC039353.1 | - | 1 | 1006 |
Gene Symbol: TTLL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AI369631.1 | - | 1 | 74 |