Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NUP93 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK125165.1 | - | 1 | 1359 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | DA513202.1 | - | 1 | 233 | Chr. 16 - 56845842 - 56846400 [+] on Build GRCh38 |