Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MTMR9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK022071.1 | - | 1 | 601 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AU120884.1 | - | 1 | 601 | Chr. 8 - 11300086 - 11300812 [+] on Build GRCh38 |