Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HNF4A-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC071794.1 | - | 2, 3 | 559 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BG427859.1 | - | 560 | Chr. - - [-] on Build |