Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BNIP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001320674.1 | NP_001307603.1 | 11 | 4822 | ||
NM_001320675.1 | NP_001307604.1 | 10 | 4581 | |||
NM_004330.3 | NP_004321.2 | 10 | 4786 | |||
GenBank mRNA | AK125533.1 | - | 1 | 1117 |