Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HIVEP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127714.2 | NP_001121186.1 | 8 | 10597 | ||
NM_024503.4 | NP_078779.2 | 9 | 10680 | |||
XM_011541884.2 | XP_011540186.1 | 10 | 10612 | |||
XM_017001992.1 | XP_016857481.1 | 10 | 11859 | |||
XM_017001993.1 | XP_016857482.1 | 8 | 10812 | |||
XM_017001994.1 | XP_016857483.1 | 9 | 10889 | |||
GenBank mRNA | BC080552.1 | - | 1 | 293 |