Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ELP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001242875.2 | NP_001229804.1 | 23 | 5861 | ||
NM_001242876.2 | NP_001229805.1 | 21 | 5651 | |||
NM_001242877.2 | NP_001229806.1 | 21 | 5588 | |||
NM_001242878.2 | NP_001229807.1 | 20 | 5456 | |||
NM_001242879.2 | NP_001229808.1 | 19 | 5456 | |||
NM_001324465.1 | NP_001311394.1 | 21 | 5534 | |||
NM_001324466.1 | NP_001311395.1 | 22 | 5783 | |||
NM_001324467.1 | NP_001311396.1 | 21 | 5516 | |||
NM_001324468.1 | NP_001311397.1 | 21 | 5587 | |||
NM_018255.3 | NP_060725.1 | 22 | 5666 | |||
NR_040110.2 | 22 | 5614 | ||||
NR_136897.1 | 21 | 5547 | ||||
NR_136898.1 | 22 | 5611 | ||||
NR_137173.1 | 21 | 5532 | ||||
GenBank mRNA | AK095780.1 | - | 1 | 2505 |