Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ITSN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_005261026.2 | XP_005261083.1 | 39 | 16881 | ||
XM_017028428.1 | XP_016883917.1 | 41 | 16816 | |||
XM_017028429.1 | XP_016883918.1 | 40 | 16785 | |||
XM_017028430.1 | XP_016883919.1 | 39 | 16770 | |||
XM_017028431.1 | XP_016883920.1 | 39 | 16728 | |||
XM_017028432.1 | XP_016883921.1 | 38 | 16713 | |||
XM_017028433.1 | XP_016883922.1 | 38 | 16683 | |||
XM_017028434.1 | XP_016883923.1 | 37 | 16668 | |||
XM_017028435.1 | XP_016883924.1 | 38 | 16572 | |||
XM_017028436.1 | XP_016883925.1 | 37 | 16557 | |||
XM_017028437.1 | XP_016883926.1 | 35 | 16236 | |||
GenBank mRNA | AK056104.1 | - | 1 | 2078 | ||
GU245887.1 | - | 1 | 308 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BM930168.1 | - | 1 | 488 | Chr. 21 - 33899197 - 33899759 [+] on Build GRCh38 |