Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ITSN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_005261026.2 | XP_005261083.1 | 39 | 16131 | ||
XM_017028428.1 | XP_016883917.1 | 41 | 16066 | |||
XM_017028429.1 | XP_016883918.1 | 40 | 16035 | |||
XM_017028430.1 | XP_016883919.1 | 39 | 16020 | |||
XM_017028431.1 | XP_016883920.1 | 39 | 15978 | |||
XM_017028432.1 | XP_016883921.1 | 38 | 15963 | |||
XM_017028433.1 | XP_016883922.1 | 38 | 15933 | |||
XM_017028434.1 | XP_016883923.1 | 37 | 15918 | |||
XM_017028435.1 | XP_016883924.1 | 38 | 15822 | |||
XM_017028436.1 | XP_016883925.1 | 37 | 15807 | |||
XM_017028437.1 | XP_016883926.1 | 35 | 15486 | |||
GenBank mRNA | AK056104.1 | - | 1 | 1328 |