Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TEF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AB051442.1 | - | 1 | 2540 |
Gene Symbol: LOC100996598 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017029163.1 | XP_016884652.1 | 2 | 245 |