Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TSC22D1-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_038381.1 | 3 | 890 | |||
GenBank mRNA | BC036219.1 | - | 1 | 991 | ||
BC040675.2 | - | 3 | 893 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BU570099.1 | - | 1 | 328 | Chr. 13 - 44578153 - 44578911 [+] on Build GRCh38 |