Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAN1A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005907.3 | NP_005898.2 | 13 | 4711 | ||
XM_005266986.4 | XP_005267043.1 | 13 | 4567 | |||
XM_011535833.2 | XP_011534135.1 | 12 | 3763 | |||
GenBank mRNA | AK024588.1 | - | 1 | 936 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BG565453.1 | - | 1 | 544 | Chr. 6 - 119177427 - 119178059 [-] on Build GRCh38 |