Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAN1A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005907.3 | NP_005898.2 | 13 | 4597 | ||
XM_005266986.4 | XP_005267043.1 | 13 | 4453 | |||
XM_011535833.2 | XP_011534135.1 | 12 | 3649 | |||
GenBank mRNA | AK024588.1 | - | 1 | 822 |