Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COTL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK125645.1 | - | 3177 |
Gene Symbol: RBM7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286045.1 | NP_001272974.1 | 5 | 3035 | ||
NM_001286046.1 | NP_001272975.1 | 5 | 3031 | |||
NM_001286047.1 | NP_001272976.1 | 4 | 2872 | |||
NM_001286048.1 | NP_001272977.1 | 3 | 2784 | |||
NM_016090.3 | NP_057174.1 | 5 | 3032 | |||
GenBank mRNA | AB209753.1 | - | 4 | 4582 |