Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SYT16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006720272.3 | XP_006720335.1 | 8 | 14109 | ||
XM_011537222.2 | XP_011535524.1 | 8 | 14017 | |||
XM_011537223.2 | XP_011535525.1 | 7 | 13223 | |||
XM_011537224.2 | XP_011535526.1 | 8 | 13369 | |||
XM_011537225.2 | XP_011535527.1 | 8 | 13668 | |||
XM_011537227.2 | XP_011535529.2 | 6 | 13046 | |||
XM_011537228.2 | XP_011535530.1 | 6 | 12989 | |||
XM_011537229.2 | XP_011535531.1 | 5 | 12833 | |||
XM_017021693.1 | XP_016877182.1 | 9 | 4785 | |||
XM_017021694.1 | XP_016877183.1 | 8 | 13875 | |||
XM_017021695.1 | XP_016877184.1 | 9 | 14020 | |||
XM_017021696.1 | XP_016877185.1 | 7 | 13168 | |||
XM_017021697.1 | XP_016877186.1 | 7 | 13007 | |||
XM_017021698.1 | XP_016877187.1 | 7 | 13292 | |||
GenBank mRNA | AK055647.1 | - | 1 | 1347 |