Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SYT16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006720272.3 | XP_006720335.1 | 8 | 13297 | ||
XM_011537222.2 | XP_011535524.1 | 8 | 13205 | |||
XM_011537223.2 | XP_011535525.1 | 7 | 12411 | |||
XM_011537224.2 | XP_011535526.1 | 8 | 12557 | |||
XM_011537225.2 | XP_011535527.1 | 8 | 12856 | |||
XM_011537227.2 | XP_011535529.2 | 6 | 12234 | |||
XM_011537228.2 | XP_011535530.1 | 6 | 12177 | |||
XM_011537229.2 | XP_011535531.1 | 5 | 12021 | |||
XM_017021693.1 | XP_016877182.1 | 9 | 3973 | |||
XM_017021694.1 | XP_016877183.1 | 8 | 13063 | |||
XM_017021695.1 | XP_016877184.1 | 9 | 13208 | |||
XM_017021696.1 | XP_016877185.1 | 7 | 12356 | |||
XM_017021697.1 | XP_016877186.1 | 7 | 12195 | |||
XM_017021698.1 | XP_016877187.1 | 7 | 12480 | |||
GenBank mRNA | AK055647.1 | - | 1 | 535 |