Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BBC3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK124984.1 | - | 1 | 516 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | DA378578.1 | - | 1 | 518 | Chr. 19 - 47222006 - 47222568 [-] on Build GRCh38 |