Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM178B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001195278.1 | NP_001182207.1 | 4 | 5390 | ||
GenBank mRNA | AK096481.1 | - | 1 | 2736 |