Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DLGAP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001003809.2 | NP_001003809.1 | 10 | 3964 | ||
NM_001242762.1 | NP_001229691.1 | 10 | 3907 | |||
NM_001242763.1 | NP_001229692.1 | 11 | 4021 | |||
NM_001242764.1 | NP_001229693.1 | 10 | 3937 | |||
NM_001242766.1 | NP_001229695.1 | 11 | 3994 | |||
NM_001308390.1 | NP_001295319.1 | 11 | 4019 | |||
NM_004746.3 | NP_004737.2 | 13 | 5321 | |||
XM_005258171.2 | XP_005258228.1 | 14 | 5392 | |||
XM_005258172.1 | XP_005258229.1 | 14 | 5387 | |||
XM_005258173.3 | XP_005258230.2 | 12 | 4183 | |||
XM_005258174.4 | XP_005258231.1 | 11 | 4240 | |||
XM_006722367.3 | XP_006722430.1 | 12 | 4351 | |||
XM_017026080.1 | XP_016881569.1 | 11 | 4320 | |||
XM_017026081.1 | XP_016881570.1 | 10 | 3984 | |||
XM_017026083.1 | XP_016881572.1 | 10 | 4209 | |||
GenBank mRNA | AK095761.1 | - | 1 | 1527 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BI489952.1 | - | 1 | 130 | Chr. 18 - 3496793 - 3497453 [-] on Build GRCh38 |