Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC102724765 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC070382.1 | - | 1 | 38 |
Gene Symbol: FAM65B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286446.1 | NP_001273375.1 | 1 | 36 | ||
XM_006715275.2 | XP_006715338.1 | 1 | 150 | |||
XM_006715281.3 | XP_006715344.1 | 1 | 150 | |||
XM_011515007.1 | XP_011513309.1 | 1 | 156 | |||
XM_011515012.1 | XP_011513314.1 | 1 | 150 | |||
GenBank mRNA | DC406827.1 | - | 1 | 36 |