Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM78B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001320302.1 | NP_001307231.1 | 2 | 2035 | ||
NR_135199.1 | 3 | 2967 | ||||
XM_017000417.1 | XP_016855906.1 | 3 | 7173 | |||
XM_017000418.1 | XP_016855907.1 | 3 | 2515 | |||
GenBank mRNA | AB593134.1 | - | 2 | 2035 | ||
AK055347.1 | - | 1 | 74 |