Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM78B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001320302.1 | NP_001307231.1 | 2 | 2473 | ||
NR_135199.1 | 3 | 3405 | ||||
XM_017000417.1 | XP_016855906.1 | 3 | 7611 | |||
XM_017000418.1 | XP_016855907.1 | 3 | 2953 | |||
GenBank mRNA | AB593134.1 | - | 2 | 2473 | ||
AK055347.1 | - | 1 | 512 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | CK430134.1 | - | 1 | 159 | Chr. 1 - 166058484 - 166059119 [-] on Build GRCh38 |