Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EBF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017009192.1 | XP_016864681.1 | 16 | 3700 | ||
XM_017009193.1 | XP_016864682.1 | 16 | 3685 | |||
XM_017009194.1 | XP_016864683.1 | 16 | 3676 | |||
XM_017009195.1 | XP_016864684.1 | 16 | 3673 | |||
XM_017009196.1 | XP_016864685.1 | 16 | 4635 | |||
XM_017009197.1 | XP_016864686.1 | 16 | 4138 | |||
XM_017009199.1 | XP_016864688.1 | 11 | 4051 | |||
XM_017009200.1 | XP_016864689.1 | 11 | 21187 | |||
XM_017009204.1 | XP_016864693.1 | 8 | 3577 | |||
GenBank mRNA | AK130895.1 | - | 1 | 1062 |