Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C14orf37 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017021029.1 | XP_016876518.1 | 8 | 13300 | ||
XM_017021030.1 | XP_016876519.1 | 8 | 13440 | |||
GenBank mRNA | BC031314.1 | - | 1 | 693 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | DB551414.1 | - | 1 | 206 | Chr. 14 - 57993547 - 57994044 [-] on Build GRCh38 |